This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DOCK5 SIRNA Assays ›
Gene Symbol:
Dock5
Gene Name:
dedicator of cytokinesis 5
Gene Aliases:
1110060D06Rik, AI666732, AI956923, BC016533, E130320D18, lr2, rlc
Chromosome Location:
Chr.14: 67751928 - 67933581 on Build GRCm38
UniGene ID:
Mm.258155
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95045

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dock5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177780.3 NP_808448.2 5 513
GenBank mRNA AK053909.1 - 5 384
AK147975.1 - 5 374
BC158046.1 - 5 373 9007351
BC158135.1 - 5 373 9088200
BC158138.1 - 5 373 9088202

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline