This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DACT2 SIRNA Assays ›
Gene Symbol:
Dact2
Gene Name:
dapper homolog 2, antagonist of beta-catenin (xenopus)
Gene Aliases:
2900084M21Rik, A630024E20, Dpr2, Frd2, dapper2
Chromosome Location:
Chr.17: 14195230 - 14204338 on Build GRCm38
UniGene ID:
Mm.79993
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95155

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dact2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172826.3 NP_766414.3 4 2619
GenBank mRNA AK041604.1 - 4 2521
AK082856.1 - 4 2538
AK141226.1 - 4 2562
BC058740.1 - 4 2547 6312377

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
epithelial cell morphogenesis
negative regulation of cell adhesion
multicellular organism development
regulation of Wnt signaling pathway
skin development
inner medullary collecting duct development
negative regulation of nodal signaling pathway
protein kinase C binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase A binding
delta-catenin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline