siRNA Details
| Gene Symbol: Haus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173400.2 | NP_775576.2 | 8 | 1047 | ||
| GenBank mRNA | AK051016.1 | - | 8 | 1068 | ||
| AK077681.1 | - | 8 | 1069 | |||
| AK169222.1 | - | 8 | 1049 | |||
| AK173206.1 | - | 8 | 1019 | |||
| BC064725.1 | - | 3 | 244 | |||
| BC068237.1 | - | 8 | 1047 | 30550558 | ||

Haus6
Genomic Map