This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HAUS6 SIRNA Assays ›
Gene Symbol:
Haus6
Gene Name:
HAUS augmin-like complex, subunit 6
Gene Aliases:
6230416J20Rik, 9830144E06, D4Ertd27e, mKIAA1574
Chromosome Location:
Chr.4: 86578855 - 86612076 on Build GRCm38
UniGene ID:
Mm.74730
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95654

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Haus6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173400.2 NP_775576.2 8, 9 1069
GenBank mRNA AK077681.1 - 8, 9 1091
AK169222.1 - 8, 9 1071
AK173206.1 - 8, 9 1041
BC064725.1 - 3, 4 266
BC068237.1 - 8, 9 1069 30550558

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spindle assembly
centrosome organization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-qpfjk:80/100.66.76.52:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline