This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXO44 SIRNA Assays ›
Gene Symbol:
Fbxo44
Gene Name:
F-box protein 44
Gene Aliases:
5730411K09, AV001623, FBG3, FBX30, Fbx6a, Fbxo6a
Chromosome Location:
Chr.4: 148152799 - 148160094 on Build GRCm38
UniGene ID:
Mm.220327
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95749

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxo44
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161851.2 NP_001155323.1 3 420
NM_001161852.2 NP_001155324.1 3 697
NM_173401.5 NP_775577.2 3 697
GenBank mRNA AK019407.1 - 3 452
AK077452.1 - 3 483
AK178346.1 - 3 412
BC028884.1 - 3 489 4165174
BM932876.1 - 2 346
BQ042735.1 - 3 480
BY252242.1 - 3 407
CX202860.1 - 2 121

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0