This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM155A SIRNA Assays ›
Gene Symbol:
Fam155a
Gene Name:
family with sequence similarity 155, member A
Gene Aliases:
6430500D16, AW121567, Tmem28
Chromosome Location:
Chr.8: 9206010 - 9773363 on Build GRCm38
UniGene ID:
Mm.268163
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95785

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam155a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173446.2 NP_775622.1 3 2810
GenBank mRNA AK032202.1 - 3 2810
AK075681.1 - 3 1941
AK083173.1 - 1 1124

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_573950.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline