This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM53C SIRNA Assays ›
Gene Symbol:
Fam53c
Gene Name:
family with sequence similarity 53, member C
Gene Aliases:
2810012G03Rik, Gm21179
Chromosome Location:
Chr.18: 34758830 - 34773760 on Build GRCm38
UniGene ID:
Mm.29424
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95965

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam53c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175104.4 NP_780313.1 5 1455
GenBank mRNA AK044469.1 - 5 1259
AK135712.1 - 4 1201
AK154214.1 - 5 1259
AK166841.1 - 4 1199
AK205243.1 - 1 106
BC044896.1 - 4 1180
BC057111.1 - 5 1455 6417613

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016605, NM_001135647, AF251040, AK310378, BC052993, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline