This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CBLN4 SIRNA Assays ›
Gene Symbol:
Cbln4
Gene Name:
cerebellin 4 precursor protein
Gene Aliases:
AI848962
Chromosome Location:
Chr.2: 172036336 - 172043466 on Build GRCm38
UniGene ID:
Mm.40555
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96979

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cbln4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175631.3 NP_783439.1 1 1377
GenBank mRNA AK032406.1 - 1 881
AK032621.1 - 1 1380
AY134663.1 - 1 209
BC094540.1 - 1 1280 5708067
BC132025.1 - 1 475 40130302
BC132027.1 - 1 475 40130304

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein secretion
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline