siRNA Details
| Gene Symbol: Pianp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145926.1 | NP_001139398.1 | 6 | 1586 | ||
| NM_001145927.1 | NP_001139399.1 | 6 | 1570 | |||
| NM_175696.4 | NP_783627.2 | 6 | 1741 | |||
| GenBank mRNA | AK082984.1 | - | 6 | 1745 | ||
| AK208418.1 | - | 1 | 311 | |||
| AK216678.1 | - | 1 | 118 | |||
| AK219084.1 | - | 1 | 102 | |||
| BC065170.1 | - | 6 | 1568 | 6413408 | ||

Pianp
Genomic Map