This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DRG2 SIRNA Assays ›
Gene Symbol:
DRG2
Gene Name:
developmentally regulated GTP binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.17: 18087867 - 18107985 on Build GRCh38
UniGene ID:
Hs.78582
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 9750

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DRG2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001388.4 NP_001379.1 2 319
XM_005256499.2 XP_005256556.1 2 301
XM_005256500.2 XP_005256557.1 2 301
XM_011523704.1 XP_011522006.1 2 301
XM_011523705.1 XP_011522007.1 2 301
XM_011523706.1 XP_011522008.1 2 214
XM_011523707.1 XP_011522009.1 2 265
XM_017024296.1 XP_016879785.1 2 214
XM_017024297.1 XP_016879786.1 2 265
GenBank mRNA AK295575.1 - 2 207
AK297569.1 - 2 214
AK313362.1 - 2 238
BC000493.2 - 2 217 2820542
BT006976.1 - 2 162
DC298961.1 - 2 319
X80754.1 - 2 209

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021354, AK012988, AJ243590, BC082564.

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
protein binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline