siRNA Details
| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291042.1 | NP_001277971.1 | 3, 4 | 217 | ||
| NM_001291043.1 | NP_001277972.1 | 3, 4 | 217 | |||
| NM_177392.3 | NP_796366.1 | 4, 5 | 289 | |||
| GenBank mRNA | AK043094.1 | - | 4, 5 | 290 | ||
| AK078505.1 | - | 3, 4 | 216 | |||
| BC110628.1 | - | 3, 4 | 217 | 40052084 | ||
| BC110629.1 | - | 3, 4 | 217 | 40052085 | ||

Tmem220
Genomic Map