This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM13103 SIRNA Assays ›
Gene Symbol:
Gm13103 Gm13088
Gene Name:
predicted gene 13103
predicted gene 13088
Gene Aliases:
E330017N17, OTTMUSG00000010433
OTTMUSG00000010332
Chromosome Location:
Chr.4: 143846497 - 143853637 on Build GRCm38
Chromosome Location:
Chr.4: 143653760 - 143660598 on Build GRCm38
UniGene ID:
Mm.252206
Mm.475072
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 98596

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm13103
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177571.3 NP_808239.2 4 1784
GenBank mRNA AK054347.1 - 4 1786
BB555205.2 - 1 583
BC096438.1 - 4 1781 30481183
Gene Symbol: Gm13088
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126325.1 NP_001119797.1 4 1771

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5wxhb:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline