This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AA986860 SIRNA Assays ›
Gene Symbol:
AA986860
Gene Name:
expressed sequence AA986860
Gene Aliases:
E030025M07, Sarg
Chromosome Location:
Chr.1: 130731976 - 130744622 on Build GRCm38
UniGene ID:
Mm.268547
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 98810

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AA986860
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177604.3 NP_808272.2 4 2546
GenBank mRNA AK028957.1 - 4 2438
AK085225.1 - 2 2843
AK086121.1 - 4 2422
BC099401.1 - 1 1485 30713499
BC117765.1 - 1 1759 40090933
BC117766.1 - 1 1759 40090934
BC129959.1 - 1 1758
BC129960.1 - 1 1759 40090940

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline