This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM9975 SIRNA Assays ›
Gene Symbol:
Gm9975
Gene Name:
predicted gene 9975
Gene Aliases:
C730049P21, ENSMUSG00000055697
Chromosome Location:
-
UniGene ID:
Mm.347746
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 98966

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm9975
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK050463.1 - 1 943
BC096608.1 - 1 1272

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline