siRNA Details
| Gene Symbol: Tgm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177911.4 | NP_808579.2 | 14 | 2319 | ||
| GenBank mRNA | AK035279.1 | - | 14 | 2320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgm4| Gene Symbol: Tgm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177911.4 | NP_808579.2 | 14 | 2319 | ||
| GenBank mRNA | AK035279.1 | - | 14 | 2320 | ||