This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SNUPN SIRNA Assays ›
Gene Symbol:
Snupn
Gene Name:
snurportin 1
Gene Aliases:
0610031A09Rik, Rnut1, Snupn1
Chromosome Location:
Chr.9: 56950924 - 56983206 on Build GRCm38
UniGene ID:
Mm.220984
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 99700

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snupn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178374.3 NP_848461.1 6 583
GenBank mRNA AK002717.1 - 6 599
AK013151.1 - 6 593
AK155315.1 - 6 600
BC023317.1 - 563
BC049638.1 - 6 583 6589249
BC061258.1 - 6 588 30251661

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus
transport
snRNA import into nucleus
molecular_function
RNA binding
protein transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-lqf76:80/100.66.77.18:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline