siRNA Details
| Gene Symbol: Mfsd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178620.3 | NP_848735.1 | 13 | 1814 | ||
| GenBank mRNA | AK011216.1 | - | 1 | 139 | ||
| AK031384.1 | - | 13 | 2009 | |||
| AK032043.1 | - | 13 | 2010 | |||
| BC016209.1 | - | 14 | 1773 | |||
| BC079583.1 | - | 13 | 1841 | |||
| BC088988.1 | - | 13 | 1814 | 6418757 | ||
| BC100547.1 | - | 13 | 3890 | |||

Mfsd11
Genomic Map