This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM108 SIRNA Assays ›
Gene Symbol:
Tmem108
Gene Name:
transmembrane protein 108
Gene Aliases:
AI462967, B130017P16Rik, R74726
Chromosome Location:
Chr.9: 103482936 - 103761872 on Build GRCm38
UniGene ID:
Mm.384704
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 99937

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem108
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178638.4 NP_848753.1 4 1085
GenBank mRNA AK044989.1 - 4 1048
AK047474.1 - 4 1047
AK052330.1 - 4 1085
AK077586.1 - 4 1040
AK086266.1 - 4 1047
AK132350.1 - 3 979
AK141589.1 - 4 1051
BC052085.1 - 4 1019 6404949

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline