siRNA Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178651.3 | NP_848766.2 | 18 | 3321 | ||
| GenBank mRNA | AK033990.1 | - | 2346 | |||
| AK082853.1 | - | 18 | 3323 | |||
| BB405788.1 | - | 1 | 159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc30a9| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178651.3 | NP_848766.2 | 18 | 3321 | ||
| GenBank mRNA | AK033990.1 | - | 2346 | |||
| AK082853.1 | - | 18 | 3323 | |||
| BB405788.1 | - | 1 | 159 | |||