siRNA Details
| Gene Symbol: Coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178693.4 | NP_848808.1 | 7 | 1699 | ||
| GenBank mRNA | AK039422.1 | - | 7 | 1700 | ||
| AK163270.1 | - | 2545 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq4| Gene Symbol: Coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178693.4 | NP_848808.1 | 7 | 1699 | ||
| GenBank mRNA | AK039422.1 | - | 7 | 1700 | ||
| AK163270.1 | - | 2545 | ||||