This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other COX7C SIRNA Assays ›
Gene Symbol:
COX7C
Gene Name:
cytochrome c oxidase subunit 7C
Gene Aliases:
-
Chromosome Location:
Chr.5: 86617967 - 86620766 on Build GRCh38
UniGene ID:
Hs.430075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS102193
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: COX7C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001867.2 NP_001858.1 1 121
GenBank mRNA AK026505.1 - 1 147
AK309192.1 - 1 114
BC001005.2 - 1 121 2821167
BC007498.2 - 1 85 3030363
BT007098.1 - 1 32
X16560.1 - 1 50

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

generation of precursor metabolites and energy
mitochondrial electron transport, cytochrome c to oxygen
hydrogen ion transmembrane transport
cytochrome-c oxidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline