siRNA Details
| Gene Symbol: ENPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001977.3 | NP_001968.3 | 15 | 2512 | ||
| XM_017007877.1 | XP_016863366.1 | 12 | 1190 | |||
| GenBank mRNA | AK313412.1 | - | 16 | 2174 | ||
| AM888273.1 | - | 15 | 2512 | |||
| AM888274.1 | - | 15 | 2512 | |||
| AM888275.1 | - | 15 | 2512 | |||
| AM888276.1 | - | 15 | 2512 | |||
| AM888277.1 | - | 15 | 2512 | |||
| AM888278.1 | - | 15 | 2512 | |||
| AM888279.1 | - | 15 | 2512 | |||
| AM888280.1 | - | 15 | 2512 | |||
| AM888281.1 | - | 15 | 2512 | |||
| AM888282.1 | - | 15 | 2512 | |||
| AM888283.1 | - | 15 | 2512 | |||
| BC094770.1 | - | 16 | 2493 | 30528477 | ||
| BX641012.1 | - | 12 | 1679 | |||
| L12468.1 | - | 16 | 2416 | |||
| L14721.1 | - | 16 | 2253 | |||

ENPEP
Genomic Map