This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FCER2 SIRNA Assays ›
Gene Symbol:
FCER2
Gene Name:
Fc fragment of IgE receptor II
Gene Aliases:
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
Chromosome Location:
Chr.19: 7688757 - 7702755 on Build GRCh38
UniGene ID:
Hs.465778
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS103598
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FCER2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207019.2 NP_001193948.2 3, 4 273
NM_001220500.1 NP_001207429.1 4, 5 409
NM_002002.4 NP_001993.2 4, 5 405
XM_005272462.4 XP_005272519.1 4, 5 1010
GenBank mRNA AK223207.1 - 4, 5 374
AK310071.1 - 4, 5 394
AK310707.1 - 4, 5 374
BC014108.2 - 4, 5 370 4309266
BC062591.1 - 4, 5 363 5216918
BC064417.1 - 3, 4 687
M14766.1 - 4, 5 377
M15059.1 - 4, 5 405
X04772.1 - 4, 5 405

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of humoral immune response mediated by circulating immunoglobulin
Notch signaling pathway
positive regulation of nitric-oxide synthase activity
positive regulation of killing of cells of other organism
positive regulation of nitric-oxide synthase biosynthetic process
integrin binding
protein binding
IgE binding
carbohydrate binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-whrt5:80/100.66.79.138:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline