This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DNAJC4 SIRNA Assays ›
Gene Symbol:
DNAJC4
Gene Name:
DnaJ heat shock protein family (Hsp40) member C4
Gene Aliases:
DANJC4, HSPF2, MCG18
Chromosome Location:
Chr.11: 64230281 - 64234281 on Build GRCh38
UniGene ID:
Hs.172847
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS105097
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DNAJC4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307980.1 NP_001294909.1 4 819
NM_001307981.1 NP_001294910.1 4 816
NM_005528.3 NP_005519.2 5 855
GenBank mRNA AF012106.1 - 2 1030
AF036874.1 - 4 462
BC032137.2 - 4 493
BC044584.1 - 5 858 4811478
BC051008.1 - 4 338
BC053947.1 - 5 858
BC057849.1 - 4 688 30349314

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to unfolded protein
protein binding
unfolded protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hn7z:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline