This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other INCENP SIRNA Assays ›
Gene Symbol:
INCENP
Gene Name:
inner centromere protein
Gene Aliases:
-
Chromosome Location:
Chr.11: 62123973 - 62155474 on Build GRCh38
UniGene ID:
Hs.142179
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS105430
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: INCENP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040694.1 NP_001035784.1 2, 3 336
NM_020238.2 NP_064623.2 2, 3 336
XM_006718533.2 XP_006718596.1 2, 3 298
XM_011544995.2 XP_011543297.1 2, 3 298
XM_011544996.2 XP_011543298.1 2, 3 298
XM_011544997.2 XP_011543299.1 2, 3 258
XM_011544998.2 XP_011543300.1 2, 3 298
GenBank mRNA AF282265.1 - 1, 2 134
AK056195.1 - 2, 3 311
AK307633.1 - 2, 3 353
AK310714.1 - 2, 3 298
AY714053.1 - 1, 2 134
BC032678.1 - 2, 3 260
BC098576.1 - 2, 3 336 30924748
CV569756.1 - 2, 3 247

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
chromosome segregation
sister chromatid cohesion
mitotic nuclear division
protein sumoylation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline