siRNA Details
| Gene Symbol: ITIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002216.2 | NP_002207.2 | 19 | 2713 | ||
| GenBank mRNA | AK225277.1 | - | 21 | 2576 | ||
| AK225329.1 | - | 19 | 2618 | |||
| AK291379.1 | - | 19 | 2661 | |||
| AK297334.1 | - | 14 | 1825 | |||
| BC132685.1 | - | 19 | 2600 | 40146707 | ||
| M18193.1 | - | 9 | 1404 | |||
| X07173.1 | - | 19 | 2614 | |||

ITIH2
Genomic Map