This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SRM SIRNA Assays ›
Gene Symbol:
SRM
Gene Name:
spermidine synthase
Gene Aliases:
PAPT, SPDSY, SPS1, SRML1
Chromosome Location:
Chr.1: 11054592 - 11060053 on Build GRCh38
UniGene ID:
Hs.76244
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS110194
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SRM
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003132.2 NP_003123.2 5 644
XM_017002179.1 XP_016857668.1 5 663
GenBank mRNA BC000309.2 - 5 610 2820975
BC033106.1 - 5 576 5420683
M34338.1 - 5 635

Back To Top

More Information


Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

polyamine metabolic process
spermidine biosynthetic process
spermidine synthase activity
protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-cfg6s:80/100.66.76.62:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0