This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF84 SIRNA Assays ›
Gene Symbol:
ZNF84
Gene Name:
zinc finger protein 84
Gene Aliases:
HPF2
Chromosome Location:
Chr.12: 133037286 - 133063304 on Build GRCh38
UniGene ID:
Hs.445019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS111562
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF84
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127372.2 NP_001120844.1 5 1149
NM_001289971.1 NP_001276900.1 5 1140
NM_001289972.1 NP_001276901.1 5 1268
NM_003428.5 NP_003419.3 5 1133
XM_005266185.1 XP_005266242.1 5 1303
XM_005266186.1 XP_005266243.1 6 1039
XM_011534832.1 XP_011533134.1 5 1083
GenBank mRNA AK314232.1 - 5 911
BC036656.1 - 5 1255 4828620
BC048350.1 - 4 717 5555056
BC062552.1 - 5 909 6536224
M27878.1 - 5 1041
X60156.2 - 4 879

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-4vr89:80/100.66.79.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0