siRNA Details
| Gene Symbol: SPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007226.1 | NP_001007227.1 | 8 | 995 | ||
| NM_001007227.1 | NP_001007228.1 | 7 | 873 | |||
| NM_001007228.1 | NP_001007229.1 | 6 | 821 | |||
| NM_001007229.1 | NP_001007230.1 | 7 | 872 | |||
| NM_001007230.1 | NP_001007231.1 | 8 | 924 | |||
| NM_003563.3 | NP_003554.1 | 7 | 943 | |||
| XM_005257723.4 | XP_005257780.1 | 7 | 1167 | |||
| XM_005257724.4 | XP_005257781.1 | 6 | 1097 | |||
| XM_017025204.1 | XP_016880693.1 | 5 | 610 | |||
| GenBank mRNA | AJ000644.1 | - | 6 | 681 | ||
| AK222589.1 | - | 6 | 646 | |||
| AK293045.1 | - | 7 | 1154 | |||
| AK293587.1 | - | 6 | 627 | |||
| AK300616.1 | - | 6 | 645 | |||
| AK303268.1 | - | 5 | 567 | |||
| AK312691.1 | - | 6 | 657 | |||
| AK316366.1 | - | 8 | 722 | |||
| BC001269.1 | - | 7 | 827 | 3461565 | ||
| BC003385.1 | - | 6 | 638 | 3445878 | ||

SPOP
Genomic Map