siRNA Details
| Gene Symbol: OSBPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278649.1 | NP_001265578.1 | 9 | 997 | ||
| NM_014835.3 | NP_055650.1 | 9 | 1016 | |||
| NM_144498.2 | NP_653081.1 | 9 | 1052 | |||
| XM_017028163.1 | XP_016883652.1 | 9 | 1052 | |||
| XM_017028164.1 | XP_016883653.1 | 10 | 1357 | |||
| XM_017028165.1 | XP_016883654.1 | 10 | 1357 | |||
| XM_017028166.1 | XP_016883655.1 | 10 | 1106 | |||
| XM_017028167.1 | XP_016883656.1 | 10 | 1120 | |||
| XM_017028168.1 | XP_016883657.1 | 9 | 1052 | |||
| XM_017028169.1 | XP_016883658.1 | 9 | 986 | |||
| XM_017028170.1 | XP_016883659.1 | 9 | 986 | |||
| GenBank mRNA | AB018315.1 | - | 9 | 922 | ||
| AF392447.1 | - | 9 | 1013 | |||
| AK000230.1 | - | 9 | 1013 | |||
| AK093592.1 | - | 3 | 1779 | |||
| AK124174.1 | - | 3 | 373 | |||
| AK291851.1 | - | 9 | 980 | |||
| AK296595.1 | - | 9 | 958 | |||
| AK315576.1 | - | 9 | 1013 | |||
| AY028168.1 | - | 9 | 922 | |||
| BC000296.2 | - | 9 | 956 | 2819652 | ||
| BC004455.1 | - | 9 | 956 | 2819652 | ||
| BC018812.2 | - | 9 | 956 | |||
| CR456722.1 | - | 8 | 811 | |||

OSBPL2
Genomic Map