This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NCAPD2 SIRNA Assays ›
Gene Symbol:
NCAPD2
Gene Name:
non-SMC condensin I complex subunit D2
Gene Aliases:
CAP-D2, CNAP1, hCAP-D2
Chromosome Location:
Chr.12: 6494132 - 6531966 on Build GRCh38
UniGene ID:
Hs.5719
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS115009
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NCAPD2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014865.3 NP_055680.3 15 1823
GenBank mRNA AK022511.1 - 15 1823
AK125155.1 - 12 1478
AK128354.1 - 15 1735
AK303310.1 - 14 1798
BC028182.1 - 15 1835 5395775
D63880.1 - 15 2576

Back To Top

More Information


Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic chromosome condensation
meiotic chromosome condensation
cell division
chromosome separation
chromatin binding
protein binding
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0