siRNA Details
| Gene Symbol: SH2D3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005490.2 | NP_005481.2 | 5 | 991 | ||
| XM_005259467.1 | XP_005259524.1 | 5 | 941 | |||
| XM_005259472.2 | XP_005259529.1 | 4 | 554 | |||
| XM_011527608.2 | XP_011525910.1 | 4 | 1358 | |||
| XM_011527609.2 | XP_011525911.1 | 4 | 855 | |||
| XM_017026123.1 | XP_016881612.1 | 5 | 941 | |||
| GenBank mRNA | AF124249.1 | - | 5 | 948 | ||
| AK225386.1 | - | 5 | 914 | |||
| AK290293.1 | - | 5 | 1067 | |||
| AK292781.1 | - | 5 | 914 | |||
| AK299406.1 | - | 4 | 564 | |||
| AK310659.1 | - | 5 | 914 | |||
| AY358406.1 | - | 5 | 948 | |||
| BC006281.2 | - | 5 | 928 | 3958342 | ||

SH2D3A
Genomic Map