This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRG4 SIRNA Assays ›
Gene Symbol:
PRG4
Gene Name:
proteoglycan 4
Gene Aliases:
CACP, HAPO, JCAP, MSF, SZP
Chromosome Location:
Chr.1: 186296273 - 186314562 on Build GRCh38
UniGene ID:
Hs.647723
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS115578
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PRG4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127708.2 NP_001121180.2 2 75
NM_001127709.2 NP_001121181.2 2 75
NM_001127710.2 NP_001121182.2 2 75
NM_001303232.1 NP_001290161.1 2 75
NM_005807.4 NP_005798.3 2 75
XM_017000002.1 XP_016855491.1 1 371
XM_017000003.1 XP_016855492.1 1 61
GenBank mRNA AK057197.1 - 1 69
AK131434.1 - 1 59
AK297718.1 - 1 69
AK301343.1 - 2 70
AK301487.1 - 1 69
AK308235.1 - 1 69
DA977744.1 - 2 75
DA995681.1 - 2 75
DC295855.1 - 2 72
DC404857.1 - 1 69
U70136.1 - 1 57

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
immune response
cell proliferation
scavenger receptor activity
polysaccharide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline