siRNA Details
| Gene Symbol: ARID3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307939.1 | NP_001294868.1 | 4 | 874 | ||
| NM_006465.3 | NP_006456.1 | 4 | 874 | |||
| GenBank mRNA | AB208830.1 | - | 4 | 793 | ||
| AF116846.1 | - | 4 | 806 | |||
| AK298716.1 | - | 4 | 799 | |||
| AK302034.1 | - | 4 | 874 | |||
| BC041792.1 | - | 4 | 802 | 5297774 | ||
| BC045545.1 | - | 4 | 848 | |||
| BC060824.1 | - | 4 | 787 | |||

ARID3B
Genomic Map