This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BLCAP SIRNA Assays ›
Gene Symbol:
BLCAP
Gene Name:
bladder cancer associated protein
Gene Aliases:
BC10
Chromosome Location:
Chr.20: 37517417 - 37527931 on Build GRCh38
UniGene ID:
Hs.472651
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS116828
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BLCAP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167820.1 NP_001161292.1 3 569
NM_001167821.1 NP_001161293.1 2 399
NM_001167822.2 NP_001161294.1 2 456
NM_001167823.1 NP_001161295.1 2 472
NM_001317074.1 NP_001304003.1 2 449
NM_001317075.1 NP_001304004.1 2 365
NM_006698.3 NP_006689.1 2 436
XM_017027599.1 XP_016883088.1 2 1349
GenBank mRNA AF053470.1 - 2 375
AK074498.1 - 4 466
AK124822.1 - 3 294
AL049288.1 - 2 382
BC047692.1 - 2 367 6059195
BC071704.1 - 2 361 5190373
BP293276.1 - 2 472
CB155706.1 - 2 406
DA198124.1 - 2 399
DA262965.1 - 2 436
DA283758.1 - 2 365
DA632207.1 - 3 518
DA770824.1 - 2 436
DA943717.1 - 2 405

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
apoptotic nuclear changes
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0