This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BLCAP SIRNA Assays ›
Gene Symbol:
BLCAP
Gene Name:
bladder cancer associated protein
Gene Aliases:
BC10
Chromosome Location:
Chr.20: 37517417 - 37527931 on Build GRCh38
UniGene ID:
Hs.472651
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS116829
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BLCAP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167820.1 NP_001161292.1 3 611
NM_001167821.1 NP_001161293.1 2 441
NM_001167822.2 NP_001161294.1 2 498
NM_001167823.1 NP_001161295.1 2 514
NM_001317074.1 NP_001304003.1 2 491
NM_001317075.1 NP_001304004.1 2 407
NM_006698.3 NP_006689.1 2 478
XM_017027599.1 XP_016883088.1 2 1391
GenBank mRNA AF053470.1 - 2 417
AK074498.1 - 4 508
AK124822.1 - 3 336
AL049288.1 - 2 424
BC047692.1 - 2 409 6059195
BC071704.1 - 2 403 5190373
BP293276.1 - 2 514
CB155706.1 - 2 448
DA198124.1 - 2 441
DA262965.1 - 2 478
DA283758.1 - 2 407
DA632207.1 - 3 560
DA770824.1 - 2 478
DA943717.1 - 2 447

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
apoptotic nuclear changes
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0