This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other TLK2 SIRNA Assays ›
Gene Symbol:
TLK2
Gene Name:
tousled like kinase 2
Gene Aliases:
HsHPK, PKU-ALPHA
Chromosome Location:
Chr.17: 62477481 - 62615481 on Build GRCh38
UniGene ID:
Hs.445078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS117032
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284333.1
NP_001271262.1
20
2162
NM_001284363.1
NP_001271292.1
17
1757
NM_006852.3
NP_006843.2
19
2099
XM_011524215.2
XP_011522517.1
18
1806
XM_011524216.2
XP_011522518.1
20
1926
XM_011524217.2
XP_011522519.1
20
2113
XM_011524218.2
XP_011522520.1
17
1740
XM_011524220.2
XP_011522522.1
17
1710
XM_011524221.2
XP_011522523.1
19
1824
XM_011524222.2
XP_011522524.1
20
2305
XM_011524223.2
XP_011522525.1
18
1967
XM_011524224.2
XP_011522526.1
18
2025
XM_011524225.2
XP_011522527.1
18
1973
XM_011524226.2
XP_011522528.1
20
2019
XM_011524227.2
XP_011522529.1
19
2073
XM_011524228.2
XP_011522530.1
20
2173
XM_011524229.2
XP_011522531.1
21
2100
XM_017024044.1
XP_016879533.1
19
1933
XM_017024045.1
XP_016879534.1
18
2041
XM_017024046.1
XP_016879535.1
19
1863
XM_017024047.1
XP_016879536.1
19
1860
XM_017024048.1
XP_016879537.1
19
2321
XM_017024049.1
XP_016879538.1
19
2274
XM_017024050.1
XP_016879539.1
17
1806
XM_017024051.1
XP_016879540.1
18
2255
XM_017024052.1
XP_016879541.1
18
2206
XM_017024053.1
XP_016879542.1
18
1878
XM_017024054.1
XP_016879543.1
17
1904
XM_017024055.1
XP_016879544.1
16
1808
XM_017024056.1
XP_016879545.1
17
1959
XM_017024057.1
XP_016879546.1
17
1773
XM_017024058.1
XP_016879547.1
19
2051
XM_017024059.1
XP_016879548.1
19
2041
XM_017024060.1
XP_016879549.1
18
1958
GenBank mRNA
AB004884.1
-
17
1735
AF162667.1
-
19
1971
AK300586.1
-
17
1757
AX775745.1
-
19
1971
BC044925.1
-
20
2069
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of chromatin assembly or disassembly
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
negative regulation of autophagy
covalent chromatin modification
peptidyl-serine phosphorylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
cellular response to gamma radiation
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-8p9zv:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline