This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other TLK2 SIRNA Assays ›
Gene Symbol:
TLK2
Gene Name:
tousled like kinase 2
Gene Aliases:
HsHPK, PKU-ALPHA
Chromosome Location:
Chr.17: 62477481 - 62615481 on Build GRCh38
UniGene ID:
Hs.445078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS117033
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284333.1
NP_001271262.1
22
2393
NM_001284363.1
NP_001271292.1
19
1988
NM_006852.3
NP_006843.2
21
2330
XM_011524215.2
XP_011522517.1
20
2037
XM_011524216.2
XP_011522518.1
22
2157
XM_011524217.2
XP_011522519.1
22
2344
XM_011524218.2
XP_011522520.1
19
1971
XM_011524220.2
XP_011522522.1
19
1941
XM_011524221.2
XP_011522523.1
21
2055
XM_011524222.2
XP_011522524.1
22
2536
XM_011524223.2
XP_011522525.1
20
2198
XM_011524224.2
XP_011522526.1
20
2256
XM_011524225.2
XP_011522527.1
20
2204
XM_011524226.2
XP_011522528.1
22
2250
XM_011524227.2
XP_011522529.1
21
2304
XM_011524228.2
XP_011522530.1
22
2404
XM_011524229.2
XP_011522531.1
23
2331
XM_017024044.1
XP_016879533.1
21
2164
XM_017024045.1
XP_016879534.1
20
2272
XM_017024046.1
XP_016879535.1
21
2094
XM_017024047.1
XP_016879536.1
21
2091
XM_017024048.1
XP_016879537.1
21
2552
XM_017024049.1
XP_016879538.1
21
2505
XM_017024050.1
XP_016879539.1
19
2037
XM_017024051.1
XP_016879540.1
20
2486
XM_017024052.1
XP_016879541.1
20
2437
XM_017024053.1
XP_016879542.1
20
2109
XM_017024054.1
XP_016879543.1
19
2135
XM_017024055.1
XP_016879544.1
18
2039
XM_017024056.1
XP_016879545.1
19
2190
XM_017024057.1
XP_016879546.1
19
2004
XM_017024058.1
XP_016879547.1
21
2282
XM_017024059.1
XP_016879548.1
21
2272
XM_017024060.1
XP_016879549.1
20
2189
GenBank mRNA
AB004884.1
-
19
1966
AF162667.1
-
21
2202
AK300586.1
-
19
1988
AX775745.1
-
21
2202
BC044925.1
-
22
2300
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001294331, NM_001112705, NM_001294329, AF045254, AK161383, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of chromatin assembly or disassembly
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
negative regulation of autophagy
covalent chromatin modification
peptidyl-serine phosphorylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
cellular response to gamma radiation
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline