This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ICMT SIRNA Assays ›
Gene Symbol:
ICMT
Gene Name:
isoprenylcysteine carboxyl methyltransferase
Gene Aliases:
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT
Chromosome Location:
Chr.1: 6221193 - 6235984 on Build GRCh38
UniGene ID:
Hs.515688
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS118802
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ICMT
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012405.3 NP_036537.1 4 683
XM_011541140.2 XP_011539442.1 3 383
GenBank mRNA AF064084.1 - 4 653
AK092890.1 - 5 624
BC017037.1 - 5 778
BC028168.1 - 4 639 5245125
CR541671.1 - 4 610
CR541711.1 - 4 610

Back To Top

More Information


Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
C-terminal protein methylation
protein targeting to membrane
post-translational protein modification
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
protein C-terminal carboxyl O-methyltransferase activity
protein C-terminal S-isoprenylcysteine carboxyl O-methyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline