This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF473 SIRNA Assays ›
Gene Symbol:
ZNF473
Gene Name:
zinc finger protein 473
Gene Aliases:
ZFP100, ZN473
Chromosome Location:
Chr.19: 50025806 - 50048776 on Build GRCh38
UniGene ID:
Hs.440553
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS119458
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF473
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017026574.1 XP_016882063.1 3 2722
GenBank mRNA AB032967.1 - 5 1479
AF454744.1 - 4 1434
AK054671.1 - 1 621
AK292452.1 - 5 1372
AK302293.1 - 4 1329
BC018612.1 - 5 1402 4634056
BX648093.1 - 5 1523

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
termination of RNA polymerase II transcription
mRNA 3'-end processing by stem-loop binding and cleavage
histone mRNA metabolic process
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-qds24:80/100.66.78.12:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0