This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PKN3 SIRNA Assays ›
Gene Symbol:
PKN3
Gene Name:
protein kinase N3
Gene Aliases:
UTDP4-1
Chromosome Location:
Chr.9: 128702497 - 128720920 on Build GRCh38
UniGene ID:
Hs.300485
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS121098
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PKN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317926.1 NP_001304855.1 5 1011
NM_013355.4 NP_037487.2 5 1011
XM_005251946.3 XP_005252003.1 5 1566
XM_006717080.2 XP_006717143.1 5 1080
XM_017014649.1 XP_016870138.1 5 1082
XM_017014650.1 XP_016870139.1 5 1220
GenBank mRNA AB019692.1 - 5 599
BC023327.1 - 5 992
BC032794.1 - 5 718
BC062558.1 - 5 614 6649806

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
signal transduction
epithelial cell migration
peptidyl-serine phosphorylation
intracellular signal transduction
protein kinase activity
protein serine/threonine kinase activity
protein kinase C activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline