siRNA Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025780.2 | NP_001020951.1 | 3 | 1129 | ||
| NM_016014.3 | NP_057098.2 | 3 | 1129 | |||
| XM_006717134.3 | XP_006717197.2 | 3 | 580 | |||
| XM_006717135.1 | XP_006717198.1 | 3 | 1117 | |||
| XM_017014787.1 | XP_016870276.1 | 3 | 736 | |||
| XM_017014788.1 | XP_016870277.1 | 4 | 1521 | |||
| XM_017014789.1 | XP_016870278.1 | 3 | 667 | |||
| GenBank mRNA | AF151825.1 | - | 3 | 813 | ||
| AK293060.1 | - | 3 | 1129 | |||
| BC038390.1 | - | 3 | 626 | 4748883 | ||

ABHD17B
Genomic Map