This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMCO3 SIRNA Assays ›
Gene Symbol:
TMCO3
Gene Name:
transmembrane and coiled-coil domains 3
Gene Aliases:
C13orf11
Chromosome Location:
Chr.13: 113490993 - 113550229 on Build GRCh38
UniGene ID:
Hs.317593
Species:
Human
siRNA ID HSS123701
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMCO3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017905.4 NP_060375.4 8, 9 1714
XM_006719969.1 XP_006720032.1 8, 9 1495
XM_011537498.2 XP_011535800.1 8, 9 1838
XM_011537502.2 XP_011535804.1 8, 9 1714
XM_011537506.2 XP_011535808.1 6, 7 1409
XM_017020629.1 XP_016876118.1 8, 9 1714
XM_017020630.1 XP_016876119.1 6, 7 1409
XM_017020631.1 XP_016876120.1 7, 8 1522
XM_017020632.1 XP_016876121.1 7, 8 1522
XM_017020633.1 XP_016876122.1 6, 7 1409
XM_017020634.1 XP_016876123.1 6, 7 1409
XM_017020635.1 XP_016876124.1 5, 6 1217
GenBank mRNA AK000630.1 - 8, 9 1611
AK074415.1 - 5, 6 740
AY358768.1 - 8, 9 1492
BC068515.1 - 8, 9 1712 4819377

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hydrogen ion transmembrane transport
solute:proton antiporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-gjjbx:80/100.66.78.233:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0