This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS123979
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
2
242
NM_001077352.1
NP_001070820.1
2
242
NM_001308044.1
NP_001294973.1
2
242
NM_018107.4
NP_060577.3
2
242
XM_011536890.2
XP_011535192.1
3
325
XM_011536892.2
XP_011535194.1
3
324
XM_011536893.2
XP_011535195.1
3
325
XM_011536894.2
XP_011535196.1
3
325
XM_011536895.2
XP_011535197.1
3
349
XM_011536896.2
XP_011535198.1
3
325
XM_011536897.2
XP_011535199.1
2
657
XM_011536900.1
XP_011535202.1
2
239
XM_011536902.1
XP_011535204.1
2
204
XM_011536904.1
XP_011535206.1
2
191
XM_011536905.1
XP_011535207.1
2
237
XM_017021398.1
XP_016876887.1
3
327
XM_017021399.1
XP_016876888.1
3
328
XM_017021400.1
XP_016876889.1
2
192
XM_017021401.1
XP_016876890.1
2
239
XM_017021402.1
XP_016876891.1
2
150
XM_017021403.1
XP_016876892.1
2
216
XM_017021404.1
XP_016876893.1
2
166
XM_017021405.1
XP_016876894.1
2
239
XM_017021406.1
XP_016876895.1
2
657
XM_017021407.1
XP_016876896.1
2
166
XM_017021408.1
XP_016876897.1
2
204
XM_017021409.1
XP_016876898.1
2
657
XM_017021410.1
XP_016876899.1
2
226
XM_017021411.1
XP_016876900.1
2
657
XM_017021412.1
XP_016876901.1
3
325
XM_017021413.1
XP_016876902.1
1
53
XM_017021414.1
XP_016876903.1
2
204
XM_017021415.1
XP_016876904.1
2
233
XM_017021417.1
XP_016876906.1
2
191
XM_017021418.1
XP_016876907.1
2
237
XM_017021420.1
XP_016876909.1
2
657
GenBank mRNA
AF087905.1
-
2
193
AF275678.1
-
2
196
AK001344.1
-
2
192
AK125593.1
-
2
204
AK225792.1
-
2
204
AK295448.1
-
2
204
AK303668.1
-
2
204
AK308055.1
-
2
204
AK315620.1
-
2
204
AL834198.1
-
2
203
BC002566.2
-
2
220
3138909
BC024208.2
-
2
179
2960467
BX161440.1
-
2
175
CK000110.1
-
2
216
CK003276.1
-
2
234
CR457257.1
-
1
43
DC363343.1
-
2
237
KU178687.1
-
1
43
KU178688.1
-
1
43
KU178689.1
-
1
43
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-57b588f4f5-5ff4v:80/100.66.79.163:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline