This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DZANK1 SIRNA Assays ›
Gene Symbol:
DZANK1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location:
Chr.20: 18383367 - 18467185 on Build GRCh38
UniGene ID:
Hs.472225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS124073
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DZANK1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099407.1 NP_001092877.1 19, 20 2235
XM_005260742.3 XP_005260799.1 19, 20 2287
XM_005260743.3 XP_005260800.1 18, 19 2141
XM_006723584.2 XP_006723647.1 17, 18 2062
XM_011529268.2 XP_011527570.1 19, 20 2405
XM_011529271.2 XP_011527573.1 19, 20 3018
XM_011529272.2 XP_011527574.1 19, 20 2401
XM_011529275.2 XP_011527577.1 18, 19 2891
XM_011529277.2 XP_011527579.1 11, 12 3132
XM_017027909.1 XP_016883398.1 19, 20 2388
XM_017027910.1 XP_016883399.1 19, 20 2271
XM_017027911.1 XP_016883400.1 19, 20 2267
XM_017027912.1 XP_016883401.1 19, 20 2974
XM_017027913.1 XP_016883402.1 19, 20 2959
XM_017027914.1 XP_016883403.1 19, 20 2213
XM_017027915.1 XP_016883404.1 19, 20 2209
XM_017027916.1 XP_016883405.1 17, 18 2773
XM_017027917.1 XP_016883406.1 20, 21 3068
XM_017027918.1 XP_016883407.1 20, 21 2322
XM_017027919.1 XP_016883408.1 20, 21 2318
XM_017027920.1 XP_016883409.1 18, 19 2277
XM_017027921.1 XP_016883410.1 12, 13 1439
GenBank mRNA AK001462.1 - 13, 14 1422
AK026306.1 - 5, 6 444
AK055544.1 - 18, 19 2030
AK131451.1 - 21, 22 3058
AK295073.1 - 19, 20 2145
AK299746.1 - 19, 20 2158
BC130341.1 - 18, 19 2063 40146372
BC144153.1 - 18, 19 2063 9052673
CR749327.1 - 20, 21 2287
DQ104738.1 - 10, 11 1156

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy in response to mitochondrial depolarization
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-grw2s:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline