This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DZANK1 SIRNA Assays ›
Gene Symbol:
DZANK1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location:
Chr.20: 18383367 - 18467185 on Build GRCh38
UniGene ID:
Hs.472225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS124074
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DZANK1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099407.1 NP_001092877.1 21 2427
XM_005260742.3 XP_005260799.1 21 2479
XM_005260743.3 XP_005260800.1 20 2333
XM_006723584.2 XP_006723647.1 19 2254
XM_011529268.2 XP_011527570.1 21 2597
XM_011529271.2 XP_011527573.1 21 3210
XM_011529272.2 XP_011527574.1 21 2593
XM_011529275.2 XP_011527577.1 20 3083
XM_011529277.2 XP_011527579.1 13 3324
XM_017027909.1 XP_016883398.1 21 2580
XM_017027910.1 XP_016883399.1 21 2463
XM_017027911.1 XP_016883400.1 21 2459
XM_017027912.1 XP_016883401.1 21 3166
XM_017027913.1 XP_016883402.1 21 3151
XM_017027914.1 XP_016883403.1 21 2405
XM_017027915.1 XP_016883404.1 21 2401
XM_017027916.1 XP_016883405.1 19 2965
XM_017027917.1 XP_016883406.1 22 3260
XM_017027918.1 XP_016883407.1 22 2514
XM_017027919.1 XP_016883408.1 22 2510
XM_017027920.1 XP_016883409.1 20 2469
XM_017027921.1 XP_016883410.1 14 1631
GenBank mRNA AK001462.1 - 15 1614
AK026306.1 - 7 636
AK055544.1 - 20 2222
AK131451.1 - 23 3250
AK295073.1 - 21 2337
AK299746.1 - 21 2350
BC130341.1 - 20 2255 40146372
BC144153.1 - 20 2255 9052673
CR749327.1 - 22 2479
DQ104738.1 - 12 1348

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy in response to mitochondrial depolarization
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline