This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FOXJ2 SIRNA Assays ›
Gene Symbol:
FOXJ2
Gene Name:
forkhead box J2
Gene Aliases:
FHX
Chromosome Location:
Chr.12: 8032680 - 8055522 on Build GRCh38
UniGene ID:
Hs.120844
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS125088
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FOXJ2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018416.2 NP_060886.1 8 2393
XM_011520760.2 XP_011519062.1 8 2476
XM_011520761.2 XP_011519063.1 9 2577
XM_017019640.1 XP_016875129.1 7 1140
GenBank mRNA AF155132.1 - 8 1797
AF155133.1 - 8 1797
BC126396.1 - 8 1363 8992112
BC136305.1 - 8 1363 9020292

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline