This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WSB2 SIRNA Assays ›
Gene Symbol:
WSB2
Gene Name:
WD repeat and SOCS box containing 2
Gene Aliases:
SBA2
Chromosome Location:
Chr.12: 118032687 - 118062430 on Build GRCh38
UniGene ID:
Hs.506985
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID HSS125208
Size 20 nmol
Availability Made To Order
Catalog # 1299001
Price 459.00
Your Price
Online offer:
459.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: WSB2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278557.1 NP_001265486.1 5 659
NM_001278558.1 NP_001265487.1 3 382
NM_018639.4 NP_061109.1 5 759
XM_017019642.1 XP_016875131.1 5 661
GenBank mRNA AF163324.1 - 4 596
AF229181.1 - 5 698
AK295555.1 - 5 660
AK298513.1 - 3 285
AK310892.1 - 5 637
BC015887.2 - 5 649 3910968

Back To Top

More Information


Gene Ontology Categories:

Function(s)

protein ubiquitination
intracellular signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline