This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other LYRM1 SIRNA Assays ›
Gene Symbol:
LYRM1
Gene Name:
LYR motif containing 1
Gene Aliases:
A211C6.1
Chromosome Location:
Chr.16: 20899626 - 20925006 on Build GRCh38
UniGene ID:
Hs.745012
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS126068
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LYRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128301.2
NP_001121773.1
4, 5
710
NM_001128302.2
NP_001121774.1
3, 4
478
NM_001302835.1
NP_001289764.1
5, 6
654
NM_001302836.1
NP_001289765.1
5, 6
806
NM_020424.4
NP_065157.1
4, 5
629
NR_126516.1
3, 4
551
NR_126520.1
2, 3
333
NR_126521.1
2, 3
319
NR_126527.1
2, 3
147
NR_126528.1
3, 4
565
XM_005255444.1
XP_005255501.1
4, 5
531
XM_011545900.2
XP_011544202.1
4, 5
293
XM_011545901.2
XP_011544203.1
4, 5
359
XM_011545902.1
XP_011544204.1
5, 6
630
XM_011545908.2
XP_011544210.1
4, 5
361
XM_017023475.1
XP_016878964.1
5, 6
1046
XM_017023477.1
XP_016878966.1
3, 4
881
XM_017023478.1
XP_016878967.1
4, 5
351
XM_017023479.1
XP_016878968.1
4, 5
788
XM_017023480.1
XP_016878969.1
6, 7
874
XM_017023481.1
XP_016878970.1
4, 5
503
XM_017023482.1
XP_016878971.1
3, 4
557
GenBank mRNA
AK308690.1
-
2, 3
398
AK311415.1
-
3, 4
553
AK311881.1
-
3, 4
345
AV714321.1
-
3, 4
324
BC017039.1
-
5, 6
654
3849248
BF575136.1
-
3, 4
367
BG541469.1
-
2, 3
182
BI547471.1
-
3, 4
396
BI825882.1
-
5, 6
653
BM546065.1
-
2, 3
283
BX096767.1
-
2, 3
147
CD237780.1
-
3, 4
348
CX163936.1
-
3, 4
355
DA572182.1
-
4, 5
595
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-nbfjs:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline