siRNA Details
| Gene Symbol: CBX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020649.2 | NP_065700.1 | 2 | 214 | ||
| GenBank mRNA | AF174482.1 | - | 2 | 189 | ||
| AF266479.2 | - | 2 | 145 | |||
| AK074560.1 | - | 2 | 214 | |||
| BC008937.2 | - | 2 | 111 | 2821280 | ||
| BC009376.2 | - | 2 | 167 | 4121509 | ||
| BC019289.1 | - | 2 | 111 | 2821280 | ||

CBX8
Genomic Map